The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318360.1 | NP_001305289.1 | 22 - 23 | 2741 | 167 |
NM_003640.4 | NP_003631.2 | 22 - 23 | 2886 | 167 | |
XM_011519136.1 | XP_011517438.1 | 22 - 23 | 2678 | 167 | |
XM_011519137.1 | XP_011517439.1 | 22 - 23 | 2452 | 167 | |
XM_017015238.1 | XP_016870727.1 | 20 - 21 | 2457 | 167 | |
GenBank mRNA | AB210033.1 | - | 22 - 23 | 2648 | 167 |
AF044195.1 | - | 22 - 23 | 2665 | 167 | |
AF153419.2 | - | 22 - 23 | 2672 | 167 | |
AK001641.1 | - | 22 - 23 | 2670 | 167 | |
AK127237.1 | - | 14 - 15 | 2301 | 167 | |
AK225798.1 | - | 9 - 10 | 970 | 167 | |
AK289962.1 | - | 22 - 23 | 2678 | 167 | |
AK304742.1 | - | 22 - 23 | 2383 | 167 | |
BC033094.1 | - | 22 - 23 | 2448 | 167 | |
CR749385.1 | - | 15 - 16 | 1713 | 167 |