The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318360.1 | NP_001305289.1 | 5 - 6 | 842 | 124 |
NM_003640.4 | NP_003631.2 | 5 - 6 | 987 | 124 | |
XM_011519136.1 | XP_011517438.1 | 5 - 6 | 779 | 124 | |
XM_011519137.1 | XP_011517439.1 | 5 - 6 | 553 | 124 | |
XM_017015238.1 | XP_016870727.1 | 5 - 6 | 746 | 124 | |
GenBank mRNA | AB210033.1 | - | 5 - 6 | 749 | 124 |
AF044195.1 | - | 5 - 6 | 766 | 124 | |
AF153419.2 | - | 5 - 6 | 773 | 124 | |
AK001641.1 | - | 5 - 6 | 771 | 124 | |
AK289962.1 | - | 5 - 6 | 779 | 124 | |
AK304742.1 | - | 5 - 6 | 484 | 124 | |
BC033094.1 | - | 5 - 6 | 549 | 124 |