Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM26
Entrez Gene ID
64062
Gene Name
RNA binding motif protein 26
Gene Aliases
ARRS2, C13orf10, PPP1R132, PRO1777, SE70-2, ZC3H17
Chromosome Location
Chr.13: 79311827 - 79406265 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286631.1 NP_001273560.1 17 - 18 2881 108
NM_001286632.1 NP_001273561.1 16 - 17 2803 108
NM_022118.4 NP_071401.3 16 - 17 2794 108
XM_005266491.4 XP_005266548.1 17 - 18 2957 108
XM_005266497.1 XP_005266554.1 17 - 18 2957 108
XM_006719857.1 XP_006719920.1 17 - 18 2957 108
XM_011535179.1 XP_011533481.1 17 - 18 2957 108
XM_011535180.2 XP_011533482.1 17 - 18 2957 108
XM_011535181.1 XP_011533483.1 17 - 18 2948 108
XM_011535182.1 XP_011533484.1 17 - 18 2942 108
XM_011535183.2 XP_011533485.1 17 - 18 2948 108
XM_011535184.1 XP_011533486.1 17 - 18 2927 108
XM_011535185.2 XP_011533487.1 17 - 18 2927 108
XM_011535186.2 XP_011533488.1 17 - 18 2919 108
XM_011535187.2 XP_011533489.1 17 - 18 2918 108
XM_011535188.2 XP_011533490.1 17 - 18 2919 108
XM_011535189.1 XP_011533491.1 16 - 17 2885 108
XM_011535190.1 XP_011533492.1 16 - 17 2876 108
XM_011535191.2 XP_011533493.1 16 - 17 2876 108
XM_011535192.2 XP_011533494.1 16 - 17 2847 108
XM_011535193.2 XP_011533495.1 16 - 17 2831 108
XM_011535195.1 XP_011533497.1 17 - 18 2655 108
XM_017020688.1 XP_016876177.1 17 - 18 2957 108
XM_017020689.1 XP_016876178.1 17 - 18 2933 108
XM_017020690.1 XP_016876179.1 17 - 18 2949 108
XM_017020691.1 XP_016876180.1 17 - 18 2940 108
XM_017020692.1 XP_016876181.1 17 - 18 2910 108
XM_017020693.1 XP_016876182.1 16 - 17 2885 108
XM_017020694.1 XP_016876183.1 16 - 17 2870 108
XM_017020695.1 XP_016876184.1 16 - 17 2861 108
XM_017020696.1 XP_016876185.1 16 - 17 2877 108
XM_017020697.1 XP_016876186.1 17 - 18 2949 108
XM_017020698.1 XP_016876187.1 16 - 17 2840 108
XM_017020699.1 XP_016876188.1 16 - 17 2862 108
XM_017020700.1 XP_016876189.1 16 - 17 2838 108
XM_017020701.1 XP_016876190.1 17 - 18 2957 108
XM_017020702.1 XP_016876191.1 17 - 18 2957 108
XM_017020703.1 XP_016876192.1 17 - 18 2927 108
GenBank mRNA AF273052.1 - 3 - 4 324 108
AK024610.1 - 1 - 2 153 108
AK027339.1 - 11 - 12 1577 108
AK027456.1 - 10 - 11 1452 108
AK302871.1 - 16 - 17 2412 108
AK302927.1 - 4 - 5 691 108
AL834343.1 - 12 - 13 1788 108
AX775885.1 - 10 - 11 1452 108
BC000791.2 - 1 - 2 179 108
BC041655.1 - 16 - 17 2361 108
BC111697.1 - 16 - 17 2364 108
BC111739.1 - 17 - 18 2443 108
BX648672.1 - 16 - 17 2361 108

View Full Product Details