Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM26
Entrez Gene ID
64062
Gene Name
RNA binding motif protein 26
Gene Aliases
ARRS2, C13orf10, PPP1R132, PRO1777, SE70-2, ZC3H17
Chromosome Location
Chr.13: 79311827 - 79406265 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286631.1 NP_001273560.1 5 - 6 1075 120
NM_001286632.1 NP_001273561.1 5 - 6 1075 120
NM_022118.4 NP_071401.3 5 - 6 1075 120
XM_005266491.4 XP_005266548.1 5 - 6 1112 120
XM_005266497.1 XP_005266554.1 5 - 6 1112 120
XM_006719857.1 XP_006719920.1 5 - 6 1112 120
XM_011535179.1 XP_011533481.1 5 - 6 1112 120
XM_011535180.2 XP_011533482.1 5 - 6 1112 120
XM_011535181.1 XP_011533483.1 5 - 6 1112 120
XM_011535182.1 XP_011533484.1 5 - 6 1112 120
XM_011535183.2 XP_011533485.1 5 - 6 1112 120
XM_011535184.1 XP_011533486.1 5 - 6 1112 120
XM_011535185.2 XP_011533487.1 5 - 6 1112 120
XM_011535186.2 XP_011533488.1 5 - 6 1119 120
XM_011535187.2 XP_011533489.1 5 - 6 1112 120
XM_011535188.2 XP_011533490.1 5 - 6 1119 120
XM_011535189.1 XP_011533491.1 5 - 6 1112 120
XM_011535190.1 XP_011533492.1 5 - 6 1112 120
XM_011535191.2 XP_011533493.1 5 - 6 1112 120
XM_011535192.2 XP_011533494.1 5 - 6 1119 120
XM_011535193.2 XP_011533495.1 5 - 6 1112 120
XM_011535195.1 XP_011533497.1 5 - 6 810 120
XM_017020688.1 XP_016876177.1 5 - 6 1112 120
XM_017020689.1 XP_016876178.1 5 - 6 1112 120
XM_017020690.1 XP_016876179.1 5 - 6 1119 120
XM_017020691.1 XP_016876180.1 5 - 6 1119 120
XM_017020692.1 XP_016876181.1 5 - 6 1119 120
XM_017020693.1 XP_016876182.1 5 - 6 1112 120
XM_017020694.1 XP_016876183.1 5 - 6 1112 120
XM_017020695.1 XP_016876184.1 5 - 6 1112 120
XM_017020696.1 XP_016876185.1 5 - 6 1119 120
XM_017020697.1 XP_016876186.1 5 - 6 1119 120
XM_017020698.1 XP_016876187.1 5 - 6 1112 120
XM_017020699.1 XP_016876188.1 5 - 6 1119 120
XM_017020700.1 XP_016876189.1 5 - 6 1119 120
XM_017020701.1 XP_016876190.1 5 - 6 1112 120
XM_017020702.1 XP_016876191.1 5 - 6 1112 120
XM_017020703.1 XP_016876192.1 5 - 6 1112 120
GenBank mRNA AK302871.1 - 5 - 6 684 120
BC041655.1 - 5 - 6 642 120
BC111697.1 - 5 - 6 636 120
BC111739.1 - 5 - 6 636 120
BX648672.1 - 5 - 6 633 120

View Full Product Details