Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM26
Entrez Gene ID
64062
Gene Name
RNA binding motif protein 26
Gene Aliases
ARRS2, C13orf10, PPP1R132, PRO1777, SE70-2, ZC3H17
Chromosome Location
Chr.13: 79311827 - 79406265 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286631.1 NP_001273560.1 6 - 7 1344 124
NM_001286632.1 NP_001273561.1 6 - 7 1344 124
NM_022118.4 NP_071401.3 6 - 7 1344 124
XM_005266491.4 XP_005266548.1 6 - 7 1381 124
XM_005266497.1 XP_005266554.1 6 - 7 1381 124
XM_006719857.1 XP_006719920.1 6 - 7 1381 124
XM_011535179.1 XP_011533481.1 6 - 7 1381 124
XM_011535180.2 XP_011533482.1 6 - 7 1381 124
XM_011535181.1 XP_011533483.1 6 - 7 1381 124
XM_011535182.1 XP_011533484.1 6 - 7 1381 124
XM_011535183.2 XP_011533485.1 6 - 7 1381 124
XM_011535184.1 XP_011533486.1 6 - 7 1381 124
XM_011535185.2 XP_011533487.1 6 - 7 1381 124
XM_011535186.2 XP_011533488.1 6 - 7 1388 124
XM_011535187.2 XP_011533489.1 6 - 7 1381 124
XM_011535188.2 XP_011533490.1 6 - 7 1388 124
XM_011535189.1 XP_011533491.1 6 - 7 1381 124
XM_011535190.1 XP_011533492.1 6 - 7 1381 124
XM_011535191.2 XP_011533493.1 6 - 7 1381 124
XM_011535192.2 XP_011533494.1 6 - 7 1388 124
XM_011535193.2 XP_011533495.1 6 - 7 1381 124
XM_011535195.1 XP_011533497.1 6 - 7 1079 124
XM_017020688.1 XP_016876177.1 6 - 7 1381 124
XM_017020689.1 XP_016876178.1 6 - 7 1381 124
XM_017020690.1 XP_016876179.1 6 - 7 1388 124
XM_017020691.1 XP_016876180.1 6 - 7 1388 124
XM_017020692.1 XP_016876181.1 6 - 7 1388 124
XM_017020693.1 XP_016876182.1 6 - 7 1381 124
XM_017020694.1 XP_016876183.1 6 - 7 1381 124
XM_017020695.1 XP_016876184.1 6 - 7 1381 124
XM_017020696.1 XP_016876185.1 6 - 7 1388 124
XM_017020697.1 XP_016876186.1 6 - 7 1388 124
XM_017020698.1 XP_016876187.1 6 - 7 1381 124
XM_017020699.1 XP_016876188.1 6 - 7 1388 124
XM_017020700.1 XP_016876189.1 6 - 7 1388 124
XM_017020701.1 XP_016876190.1 6 - 7 1381 124
XM_017020702.1 XP_016876191.1 6 - 7 1381 124
XM_017020703.1 XP_016876192.1 6 - 7 1381 124
GenBank mRNA AK027339.1 - 1 - 2 275 124
AK302871.1 - 6 - 7 953 124
AL834343.1 - 1 - 2 257 124
BC041655.1 - 6 - 7 911 124
BC111697.1 - 6 - 7 905 124
BC111739.1 - 6 - 7 905 124
BX648672.1 - 6 - 7 902 124

View Full Product Details