The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017369.2 | NP_001017369.1 | 2 - 3 | 248 | 108 |
NM_006745.4 | NP_006736.1 | 3 - 4 | 534 | 108 | |
XM_005263176.2 | XP_005263233.1 | 3 - 4 | 651 | 108 | |
GenBank mRNA | AK292418.1 | - | 3 - 4 | 479 | 108 |
AK295432.1 | - | 2 - 3 | 194 | 108 | |
AK309206.1 | - | 3 - 4 | 479 | 108 | |
BC010653.1 | - | 2 - 3 | 428 | 108 | |
BC107879.1 | - | 3 - 4 | 496 | 108 | |
BU940384.1 | - | - | 209 | 108 | |
BX441001.2 | - | 3 - 4 | 534 | 108 | |
U60205.1 | - | 2 - 3 | 430 | 108 | |
U93162.1 | - | 3 - 4 | 494 | 108 |