The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000840.2 | NP_000831.2 | 2 - 3 | 1570 | 82 |
XM_011516088.1 | XP_011514390.1 | 2 - 3 | 744 | 82 | |
XM_017012073.1 | XP_016867562.1 | 2 - 3 | 744 | 82 | |
GenBank mRNA | AK291406.1 | - | 2 - 3 | 932 | 82 |
BC022496.1 | - | 2 - 3 | 768 | 82 | |
BC041407.1 | - | 2 - 3 | 1577 | 82 | |
DQ315361.1 | - | 2 - 3 | 642 | 82 | |
EU432123.1 | - | 1 - 2 | 471 | 82 | |
X77748.1 | - | 2 - 3 | 723 | 82 |