The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018317.3 | NP_060787.2 | 4 - 5 | 559 | 114 |
XM_011513852.2 | XP_011512154.1 | 4 - 5 | 399 | 114 | |
XM_017008363.1 | XP_016863852.1 | 4 - 5 | 399 | 114 | |
XM_017008364.1 | XP_016863853.1 | 4 - 5 | 399 | 114 | |
GenBank mRNA | AK001944.1 | - | 4 - 5 | 399 | 114 |
AK225395.1 | - | 4 - 5 | 399 | 114 | |
AK289631.1 | - | 4 - 5 | 399 | 114 | |
AK310795.1 | - | 4 - 5 | 406 | 114 | |
BC031642.1 | - | 4 - 5 | 355 | 114 |