The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015641.3 | NP_056456.1 | 6 - 7 | 1285 | 58 |
NM_152829.2 | NP_690042.1 | 6 - 7 | 1119 | 58 | |
XM_005250258.1 | XP_005250315.1 | 5 - 6 | 1153 | 58 | |
GenBank mRNA | AF245356.1 | - | 6 - 7 | 1211 | 58 |
AF245357.1 | - | 6 - 7 | 1089 | 58 | |
AJ250865.1 | - | 6 - 7 | 1198 | 58 | |
AK021575.1 | - | 6 - 7 | 1229 | 58 | |
AK291802.1 | - | 6 - 7 | 1100 | 58 | |
AK300546.1 | - | 4 - 5 | 640 | 58 | |
AK315644.1 | - | 6 - 7 | 1197 | 58 | |
AL050162.1 | - | 2 - 3 | 217 | 58 | |
BC001451.1 | - | 6 - 7 | 1251 | 58 |