Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STX17
Entrez Gene ID
55014
Gene Name
syntaxin 17
Gene Aliases
-
Chromosome Location
Chr.9: 99906633 - 99974537 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017919.2 NP_060389.2 5 - 6 666 62
XM_011518820.2 XP_011517122.1 5 - 6 749 62
XM_011518821.2 XP_011517123.1 5 - 6 997 62
XM_011518823.2 XP_011517125.1 3 - 4 487 62
XM_017014875.1 XP_016870364.1 6 - 7 843 62
GenBank mRNA AK095262.1 - 5 - 6 1277 62
AL833620.1 - 5 - 6 1178 62
AL834371.1 - 1 - 2 103 62
BC070269.1 - 5 - 6 609 62
BC101564.1 - 4 - 5 603 62
BC101566.1 - 4 - 5 603 62
BC111009.1 - 5 - 6 626 62

View Full Product Details