Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZSCAN18
Entrez Gene ID
65982
Gene Name
zinc finger and SCAN domain containing 18
Gene Aliases
ZNF447
Chromosome Location
Chr.19: 58083834 - 58118433 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145542.1 NP_001139014.1 4 - 5 838 77
NM_001145543.1 NP_001139015.1 4 - 5 949 77
NM_001145544.1 NP_001139016.1 3 - 4 486 77
NM_023926.4 NP_076415.3 4 - 5 1193 77
XM_005259174.4 XP_005259231.1 4 - 5 836 77
XM_006723335.2 XP_006723398.1 4 - 5 890 77
XM_011527237.2 XP_011525539.1 4 - 5 845 77
XM_011527238.1 XP_011525540.1 5 - 6 1044 77
XM_011527239.2 XP_011525541.1 5 - 6 1016 77
XM_017027169.1 XP_016882658.1 4 - 5 949 77
XM_017027170.1 XP_016882659.1 4 - 5 890 77
XM_017027171.1 XP_016882660.1 4 - 5 837 77
GenBank mRNA AK022957.1 - 4 - 5 835 77
AK126140.1 - 1 - 2 711 77
AK225548.1 - 4 - 5 807 77
AK294376.1 - 4 - 5 808 77
AY280799.1 - 4 - 5 842 77
BC006205.2 - 3 - 4 334 77
BC022846.2 - 4 - 5 1040 77
DA611622.1 - 3 - 4 351 77

View Full Product Details