The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271899.1 | NP_001258828.1 | 4 - 5 | 600 | 73 |
NM_001271900.1 | NP_001258829.1 | 5 - 6 | 794 | 73 | |
NM_001271948.1 | NP_001258877.1 | 4 - 5 | 1127 | 73 | |
NM_181674.2 | NP_858060.2 | 4 - 5 | 925 | 73 | |
NM_181675.3 | NP_858061.2 | 4 - 5 | 647 | 73 | |
NM_181676.2 | NP_858062.1 | 3 - 4 | 731 | 73 | |
NM_181677.2 | NP_858063.1 | 5 - 6 | 557 | 73 | |
NM_181678.2 | NP_858064.1 | 5 - 6 | 570 | 73 | |
NR_073526.1 | 3 - 4 | 1042 | 73 | ||
NR_073527.1 | 4 - 5 | 679 | 73 | ||
GenBank mRNA | AK056192.1 | - | 4 - 5 | 607 | 73 |
AK289717.1 | - | 4 - 5 | 576 | 73 | |
AK294659.1 | - | 5 - 6 | 703 | 73 | |
AK314810.1 | - | 4 - 5 | 924 | 73 | |
AL540315.3 | - | 2 - 3 | 260 | 73 | |
BC031790.1 | - | 4 - 5 | 815 | 73 | |
BI669304.1 | - | 5 - 6 | 555 | 73 | |
BX647887.1 | - | 4 - 5 | 660 | 73 | |
M64930.1 | - | 3 - 4 | 853 | 73 |