The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030571.3 | NP_085048.1 | 5 - 6 | 707 | 97 |
GenBank mRNA | AB097010.1 | - | 5 - 6 | 643 | 97 |
AB097037.1 | - | 5 - 6 | 643 | 97 | |
AK075524.1 | - | 5 - 6 | 646 | 97 | |
AK315481.1 | - | 5 - 6 | 652 | 97 | |
AL832993.1 | - | 2 - 3 | 188 | 97 | |
AY192728.1 | - | 5 - 6 | 663 | 97 | |
BC004317.1 | - | 5 - 6 | 595 | 97 |