The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301015.1 | NP_001287944.1 | 9 - 10 | 1501 | 84 |
NM_001301018.1 | NP_001287947.1 | 9 - 10 | 975 | 84 | |
NM_014945.3 | NP_055760.1 | 10 - 11 | 1147 | 84 | |
XM_005268392.1 | XP_005268449.1 | 10 - 11 | 1128 | 84 | |
XM_005268393.1 | XP_005268450.1 | 10 - 11 | 1128 | 84 | |
XM_011537609.2 | XP_011535911.1 | 10 - 11 | 1128 | 84 | |
GenBank mRNA | AB020650.1 | - | 10 - 11 | 1060 | 84 |
AK225769.1 | - | 9 - 10 | 1369 | 84 | |
AK289736.1 | - | 9 - 10 | 914 | 84 | |
DQ413174.1 | - | 9 - 10 | 937 | 84 |