The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301015.1 | NP_001287944.1 | 18 - 19 | 2335 | 57 |
NM_001301018.1 | NP_001287947.1 | 17 - 18 | 1710 | 57 | |
NM_001301027.1 | NP_001287956.1 | 15 - 16 | 2081 | 57 | |
NM_001301028.1 | NP_001287957.1 | 14 - 15 | 2002 | 57 | |
NM_014945.3 | NP_055760.1 | 19 - 20 | 1981 | 57 | |
XM_005268392.1 | XP_005268449.1 | 18 - 19 | 1863 | 57 | |
XM_005268393.1 | XP_005268450.1 | 17 - 18 | 1815 | 57 | |
XM_005268394.1 | XP_005268451.1 | 17 - 18 | 1776 | 57 | |
XM_005268395.1 | XP_005268452.1 | 16 - 17 | 1677 | 57 | |
XM_005268396.1 | XP_005268453.1 | 15 - 16 | 1629 | 57 | |
XM_017009238.1 | XP_016864727.1 | 15 - 16 | 1771 | 57 | |
GenBank mRNA | AB020650.1 | - | 19 - 20 | 1894 | 57 |
AB075881.1 | - | 4 - 5 | 1904 | 57 | |
AK225769.1 | - | 18 - 19 | 2203 | 57 | |
AK289736.1 | - | 18 - 19 | 1748 | 57 | |
AL833021.1 | - | 12 - 13 | 1089 | 57 | |
BC001665.2 | - | 14 - 15 | 1664 | 57 | |
DQ413174.1 | - | 17 - 18 | 1672 | 57 |