The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282875.1 | NP_001269804.1 | 12 - 13 | 1756 | 124 |
XM_006724782.2 | XP_006724845.1 | 12 - 13 | 1815 | 124 | |
XM_017029750.1 | XP_016885239.1 | 12 - 13 | 1815 | 124 | |
XM_017029751.1 | XP_016885240.1 | 12 - 13 | 1815 | 124 | |
GenBank mRNA | BC051825.1 | - | 11 - 12 | 1433 | 124 |
EU636009.1 | - | 12 - 13 | 1715 | 124 |