The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282874.1 | NP_001269803.1 | 24 - 25 | 3280 | 85 |
NM_001282875.1 | NP_001269804.1 | 23 - 24 | 3244 | 85 | |
GenBank mRNA | AK123541.1 | - | 1 - 2 | 1738 | 85 |
BC143954.1 | - | 24 - 25 | 3180 | 85 |