The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000358.2 | NP_000349.1 | 9 - 10 | 1429 | 63 |
GenBank mRNA | AB209598.1 | - | 3 - 4 | 4191 | 63 |
AK093916.1 | - | 9 - 10 | 1284 | 63 | |
AK094055.1 | - | 10 - 11 | 1360 | 63 | |
AK094581.1 | - | 9 - 10 | 1373 | 63 | |
AK222833.1 | - | 9 - 10 | 1337 | 63 | |
AK298603.1 | - | 4 - 5 | 649 | 63 | |
AK303643.1 | - | 9 - 10 | 1661 | 63 | |
BC000097.2 | - | 9 - 10 | 1322 | 63 | |
BC004972.1 | - | 9 - 10 | 1317 | 63 | |
BT009820.1 | - | 9 - 10 | 1268 | 63 | |
KU178357.1 | - | 9 - 10 | 1268 | 63 | |
KU178358.1 | - | 7 - 8 | 782 | 63 | |
M77349.1 | - | 9 - 10 | 1315 | 63 |