The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252020.1 | NP_001238949.1 | 23 - 24 | 3513 | 76 |
NM_001252024.1 | NP_001238953.1 | 24 - 25 | 3301 | 76 | |
NM_002420.5 | NP_002411.3 | 23 - 24 | 3215 | 76 | |
GenBank mRNA | AB115498.1 | - | 20 - 21 | 2872 | 76 |
AB115499.1 | - | 20 - 21 | 2872 | 76 | |
AB115501.1 | - | 20 - 21 | 2872 | 76 | |
AB115502.1 | - | 20 - 21 | 2854 | 76 | |
AF071787.1 | - | 23 - 24 | 3192 | 76 | |
HM135790.1 | - | 24 - 25 | 3296 | 76 | |
HM135791.1 | - | 23 - 24 | 3513 | 76 |