The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000246.3 | NP_000237.2 | 19 - 20 | 3551 | 70 |
NM_001286402.1 | NP_001273331.1 | 19 - 20 | 3554 | 70 | |
NM_001286403.1 | NP_001273332.1 | 17 - 18 | 1799 | 70 | |
NR_104444.1 | 17 - 18 | 1741 | 70 | ||
XM_011522484.2 | XP_011520786.1 | 19 - 20 | 3732 | 70 | |
GenBank mRNA | AF410154.1 | - | 19 - 20 | 4432 | 70 |
U18259.1 | - | 19 - 20 | 3556 | 70 | |
X74301.1 | - | 19 - 20 | 3533 | 70 |