The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079846.1 | NP_001073315.1 | 8 - 9 | 2032 | 136 |
NM_004380.2 | NP_004371.2 | 9 - 10 | 2146 | 136 | |
XM_005255124.4 | XP_005255181.1 | 9 - 10 | 2753 | 136 | |
XM_005255125.4 | XP_005255182.1 | 9 - 10 | 2753 | 136 | |
XM_006720848.3 | XP_006720911.1 | 9 - 10 | 2753 | 136 | |
XM_011522381.2 | XP_011520683.1 | 8 - 9 | 1635 | 136 | |
XM_011522382.2 | XP_011520684.1 | 9 - 10 | 2753 | 136 | |
XM_017022944.1 | XP_016878433.1 | 9 - 10 | 2753 | 136 | |
GenBank mRNA | AB210043.1 | - | 8 - 9 | 2032 | 136 |
AK304771.1 | - | 6 - 7 | 920 | 136 | |
U47741.1 | - | 10 - 11 | 2140 | 136 | |
U85962.3 | - | 9 - 10 | 2760 | 136 |