The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015113.3 | NP_055928.3 | 32 - 33 | 5229 | 138 |
XM_005256561.2 | XP_005256618.1 | 32 - 33 | 5293 | 138 | |
XM_017024382.1 | XP_016879871.1 | 32 - 33 | 5293 | 138 | |
XM_017024383.1 | XP_016879872.1 | 32 - 33 | 5290 | 138 | |
XM_017024384.1 | XP_016879873.1 | 7 - 8 | 1104 | 138 | |
GenBank mRNA | AB007859.4 | - | 32 - 33 | 5229 | 138 |
AK001683.1 | - | 3 - 4 | 328 | 138 | |
AK122719.1 | - | 7 - 8 | 1323 | 138 | |
BC151836.1 | - | 32 - 33 | 5229 | 138 |