Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CASK
Entrez Gene ID
8573
Gene Name
calcium/calmodulin dependent serine protein kinase
Gene Aliases
CAGH39, CAMGUK, CMG, FGS4, LIN2, MICPCH, MRXSNA, TNRC8
Chromosome Location
Chr.X: 41514934 - 41923525 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126054.2 NP_001119526.1 16 - 17 1631 143
NM_001126055.2 NP_001119527.1 15 - 16 1613 143
NM_003688.3 NP_003679.2 16 - 17 1631 143
XM_005272686.4 XP_005272743.1 15 - 16 2104 143
XM_006724566.3 XP_006724629.1 15 - 16 2104 143
XM_011543993.2 XP_011542295.1 15 - 16 1683 143
XM_011543994.2 XP_011542296.1 15 - 16 1683 143
XM_011543995.2 XP_011542297.1 15 - 16 1683 143
XM_011543996.2 XP_011542298.1 15 - 16 1683 143
XM_011543997.2 XP_011542299.1 10 - 11 1342 143
GenBank mRNA AB039327.2 - 4 - 5 789 143
AB208859.1 - 16 - 17 1575 143
AF032119.1 - 16 - 17 1612 143
AF035582.1 - 15 - 16 1582 143
AF262404.1 - 11 - 12 1071 143
AF262405.1 - 14 - 15 1489 143
AK313399.1 - 15 - 16 1766 143
AY705392.1 - 16 - 17 1591 143
BC117311.1 - 16 - 17 1604 143
BC143454.1 - 16 - 17 1604 143
BC143456.1 - 15 - 16 1586 143

View Full Product Details