Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLOD4
Entrez Gene ID
51031
Gene Name
glyoxalase domain containing 4
Gene Aliases
C17orf25, CGI-150, HC71
Chromosome Location
Chr.17: 759309 - 782331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006721540.2 XP_006721603.1 2 - 3 236 72
XM_017024711.1 XP_016880200.1 2 - 3 538 72
GenBank mRNA AF177342.1 - 3 - 4 207 72
AK296584.1 - 3 - 4 404 72

View Full Product Details