The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242466.1 | NP_001229395.1 | 3 - 4 | 441 | 145 |
NM_181504.3 | NP_852556.2 | 4 - 5 | 578 | 145 | |
NM_181523.2 | NP_852664.1 | 10 - 11 | 1878 | 145 | |
NM_181524.1 | NP_852665.1 | 4 - 5 | 710 | 145 | |
XM_005248542.3 | XP_005248599.1 | 10 - 11 | 1901 | 145 | |
XM_011543493.2 | XP_011541795.1 | 9 - 10 | 1531 | 145 | |
XM_017009585.1 | XP_016865074.1 | 10 - 11 | 2670 | 145 | |
XM_017009586.1 | XP_016865075.1 | 9 - 10 | 1491 | 145 | |
GenBank mRNA | AF279367.1 | - | 4 - 5 | 710 | 145 |
AK126345.1 | - | 3 - 4 | 1841 | 145 | |
AK223613.1 | - | 4 - 5 | 558 | 145 | |
AK294919.1 | - | 2 - 3 | 697 | 145 | |
AK299743.1 | - | 8 - 9 | 907 | 145 | |
BC030815.1 | - | 4 - 5 | 550 | 145 | |
BC094795.1 | - | 10 - 11 | 1858 | 145 | |
JX133164.1 | - | 3 - 4 | 300 | 145 | |
JX133165.1 | - | 3 - 4 | 300 | 145 | |
M61906.1 | - | 9 - 10 | 1340 | 145 |