The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004429.4 | NP_004420.1 | 2 - 3 | 1187 | 80 |
GenBank mRNA | BC016649.1 | - | 2 - 3 | 1187 | 80 |
BC052979.1 | - | 2 - 3 | 1206 | 80 | |
L37361.1 | - | 2 - 3 | 933 | 80 | |
U09303.1 | - | 2 - 3 | 1108 | 80 | |
U09304.1 | - | 2 - 3 | 714 | 80 |