The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042388.2 | NP_001035847.1 | 9 - 10 | 1042 | 88 |
NM_005134.3 | NP_005125.1 | 9 - 10 | 991 | 88 | |
NR_052003.1 | 8 - 9 | 884 | 88 | ||
XM_011525775.2 | XP_011524077.1 | 9 - 10 | 1168 | 88 | |
XM_011525776.1 | XP_011524078.1 | 9 - 10 | 1032 | 88 | |
XM_017026106.1 | XP_016881595.1 | 8 - 9 | 846 | 88 | |
XM_017026107.1 | XP_016881596.1 | 8 - 9 | 861 | 88 | |
GenBank mRNA | AF111106.1 | - | 9 - 10 | 961 | 88 |
AF200478.1 | - | 9 - 10 | 941 | 88 | |
AK027064.1 | - | 9 - 10 | 993 | 88 | |
AK292538.1 | - | 9 - 10 | 922 | 88 | |
AK293869.1 | - | 8 - 9 | 782 | 88 | |
AK315106.1 | - | 9 - 10 | 949 | 88 | |
BC060829.1 | - | 9 - 10 | 943 | 88 |