Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC25
Entrez Gene ID
548645
Gene Name
DnaJ heat shock protein family (Hsp40) member C25
Gene Aliases
bA16L21.2.1
Chromosome Location
Chr.9: 111631352 - 111654351 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015882.2 NP_001015882.2 1 - 2 394 86
GenBank mRNA BC048318.1 - 1 - 2 365 86
DR004317.1 - 1 - 2 401 86

View Full Product Details