Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAPK15
Entrez Gene ID
225689
Gene Name
mitogen-activated protein kinase 15
Gene Aliases
ERK7, ERK8
Chromosome Location
Chr.8: 143716305 - 143722463 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139021.2 NP_620590.2 1 - 2 104 72
XM_011516926.2 XP_011515228.1 1 - 2 136 72
GenBank mRNA AY065978.1 - 1 - 2 96 72
AY994058.1 - 1 - 2 63 72
BC028034.1 - 1 - 2 101 72
BP357805.1 - 1 - 2 104 72

View Full Product Details