Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP2B6
- Entrez Gene ID
-
1555
- Gene Name
- cytochrome P450 family 2 subfamily B member 6
- Gene Aliases
-
CPB6, CYP2B, CYP2B7, CYP2B7P, CYPIIB6, EFVM, IIB1, P450
- Chromosome Location
-
Chr.19: 40991200 - 41018402 on Build GRCh38
- Gene Symbol
- CYP2B7P
- Entrez Gene ID
-
1556
- Gene Name
- cytochrome P450 family 2 subfamily B member 7, pseudogene
- Gene Aliases
-
CYP2B, CYP2B7, CYP2B7P1
- Chromosome Location
-
Chr.19: 40924265 - 40950660 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_001278.1 |
|
5 - 6 |
827 |
72 |
GenBank mRNA |
DQ198366.1 |
- |
5 - 6 |
819 |
72 |
DQ198367.1 |
- |
5 - 6 |
819 |
72 |
View Full Product Details