The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011525918.2 | XP_011524220.1 | 2 - 3 | 2684 | 94 |
XM_011525920.2 | XP_011524222.1 | 2 - 3 | 124 | 94 | |
XM_017025704.1 | XP_016881193.1 | 1 - 2 | 523 | 94 | |
XM_017025705.1 | XP_016881194.1 | 3 - 4 | 2566 | 94 |