Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RUSC1
Entrez Gene ID
23623
Gene Name
RUN and SH3 domain containing 1
Gene Aliases
NESCA
Chromosome Location
Chr.1: 155320849 - 155331118 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105203.1 NP_001098673.1 3 - 4 1689 105
NM_001105204.1 NP_001098674.1 3 - 4 1689 105
NM_001105205.1 NP_001098675.1 2 - 3 711 105
NM_001278227.1 NP_001265156.1 2 - 3 754 105
NM_001278228.1 NP_001265157.1 2 - 3 754 105
NM_001278229.1 NP_001265158.1 2 - 3 754 105
NM_001278230.1 NP_001265159.1 1 - 2 553 105
NM_014328.4 NP_055143.2 2 - 3 754 105
XM_006711254.1 XP_006711317.1 4 - 5 2100 105
XM_006711256.1 XP_006711319.1 4 - 5 2100 105
XM_006711257.1 XP_006711320.1 2 - 3 368 105
XM_017000891.1 XP_016856380.1 3 - 4 2270 105
XM_017000892.1 XP_016856381.1 2 - 3 368 105
GenBank mRNA AB026894.2 - 2 - 3 178 105
AK055451.1 - 2 - 3 949 105
AK074982.1 - 2 - 3 173 105
AK093777.1 - 2 - 3 207 105
AK125378.1 - 3 - 4 1641 105
AK298730.1 - 2 - 3 264 105
AK307736.1 - 2 - 3 204 105
AK314559.1 - 2 - 3 423 105
BC001045.2 - 2 - 3 195 105
BC052277.1 - 2 - 3 711 105
BX402179.2 - 2 - 3 245 105
BX640612.1 - 1 - 2 371 105
CN305634.1 - 1 - 2 553 105
DA131200.1 - 2 - 3 264 105

View Full Product Details