The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199829.1 | NP_001186758.1 | 4 - 5 | 359 | 99 |
NM_032132.4 | NP_115508.2 | 4 - 5 | 359 | 120 | |
XM_011510053.2 | XP_011508355.1 | 4 - 5 | 427 | 120 | |
XM_011510054.2 | XP_011508356.1 | 4 - 5 | 381 | 120 | |
XM_011510055.2 | XP_011508357.1 | 3 - 4 | 177 | 120 | |
GenBank mRNA | AK097390.1 | - | 4 - 5 | 347 | 120 |
AK098633.1 | - | 4 - 5 | 317 | 120 | |
AL136755.2 | - | 4 - 5 | 356 | 99 | |
AY626344.1 | - | 4 - 5 | 356 | 99 | |
BC047406.1 | - | 4 - 5 | 347 | 120 | |
CR533505.1 | - | 3 - 4 | 241 | 99 |