The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145160.1 | NP_001138632.1 | 1 - 2 | 232 | 79 |
XM_006722865.2 | XP_006722928.1 | 1 - 2 | 232 | 79 | |
GenBank mRNA | AK023385.1 | - | 1 - 2 | 201 | 79 |
DA032511.1 | - | 1 - 2 | 232 | 79 |