Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM69
Entrez Gene ID
140691
Gene Name
tripartite motif containing 69
Gene Aliases
HSD-34, HSD34, RNF36, Trif
Chromosome Location
Chr.15: 44736362 - 44767829 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301144.1 NP_001288073.1 2 - 3 451 77
NM_001301145.1 NP_001288074.1 2 - 3 451 77
GenBank mRNA AF302088.1 - 2 - 3 299 77
AF302089.1 - 2 - 3 299 77
AL360161.1 - 2 - 3 286 77

View Full Product Details