Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM69
Entrez Gene ID
140691
Gene Name
tripartite motif containing 69
Gene Aliases
HSD-34, HSD34, RNF36, Trif
Chromosome Location
Chr.15: 44736362 - 44767829 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080745.4 NP_542783.2 2 - 3 445 70
NM_182985.4 NP_892030.3 3 - 4 922 70
GenBank mRNA AK292252.1 - 3 - 4 766 70
AY305385.1 - 3 - 4 620 70
BC024199.1 - 3 - 4 740 70
BC033314.1 - 2 - 3 241 70
BC047945.1 - 3 - 4 759 70
BM559467.1 - 3 - 4 768 70

View Full Product Details