The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145305.2 | NP_660348.2 | 1 - 2 | 381 | 112 |
XM_017029340.1 | XP_016884829.1 | 1 - 2 | 350 | 112 | |
XM_017029341.1 | XP_016884830.1 | 1 - 2 | 350 | 112 | |
GenBank mRNA | AK091304.1 | - | 1 - 2 | 296 | 112 |
AK292035.1 | - | 1 - 2 | 295 | 112 | |
AK303932.1 | - | 1 - 2 | 296 | 112 | |
BC019584.1 | - | 1 - 2 | 616 | 112 | |
BC071871.1 | - | 1 - 2 | 343 | 112 |