Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A43
Entrez Gene ID
203427
Gene Name
solute carrier family 25 member 43
Gene Aliases
-
Chromosome Location
Chr.X: 119399295 - 119454478 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145305.2 NP_660348.2 4 - 5 931 82
GenBank mRNA AK094254.1 - 5 - 6 746 82
AK292035.1 - 4 - 5 845 82
AK303932.1 - 3 - 4 673 82
BC019584.1 - 4 - 5 1166 82
BC071871.1 - 4 - 5 893 82

View Full Product Details