The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004998.3 | NP_004989.2 | 15 - 16 | 1991 | 102 |
GenBank mRNA | AB290177.1 | - | 15 - 16 | 1974 | 102 |
AK291688.1 | - | 15 - 16 | 1988 | 102 | |
BC098392.1 | - | 15 - 16 | 1990 | 102 | |
U14391.1 | - | 15 - 16 | 1994 | 102 |