The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321924.1 | NP_001308853.1 | 10 - 11 | 1619 | 79 |
NM_016213.4 | NP_057297.2 | 10 - 11 | 1541 | 79 | |
NR_135855.1 | 9 - 10 | 1414 | 79 | ||
GenBank mRNA | AK314319.1 | - | 10 - 11 | 1484 | 79 |
BC012448.1 | - | 10 - 11 | 1524 | 79 | |
CA425850.1 | - | 3 - 4 | 79 |