The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321924.1 | NP_001308853.1 | 11 - 12 | 1715 | 116 |
NM_016213.4 | NP_057297.2 | 11 - 12 | 1637 | 116 | |
NR_135855.1 | 10 - 11 | 1510 | 116 | ||
GenBank mRNA | AK314319.1 | - | 11 - 12 | 1580 | 116 |
BC012448.1 | - | 11 - 12 | 1620 | 116 | |
CA425850.1 | - | 2 - 3 | 116 |