The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052853.3 | NP_443085.2 | 3 - 4 | 1394 | 65 |
XM_006716170.3 | XP_006716233.1 | 3 - 4 | 1619 | 65 | |
GenBank mRNA | AF131745.1 | - | 3 - 4 | 1393 | 65 |
AY302592.1 | - | 3 - 4 | 1394 | 65 |