The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166135.1 | NP_001159607.1 | 3 - 4 | 495 | 81 |
NM_018077.2 | NP_060547.2 | 7 - 8 | 918 | 81 | |
XM_011516370.2 | XP_011514672.1 | 8 - 9 | 1025 | 81 | |
XM_017012389.1 | XP_016867878.1 | 8 - 9 | 1026 | 81 | |
XM_017012390.1 | XP_016867879.1 | 7 - 8 | 918 | 81 | |
GenBank mRNA | AK001239.1 | - | 7 - 8 | 918 | 81 |
AK222716.1 | - | 7 - 8 | 880 | 81 | |
AK300500.1 | - | 3 - 4 | 457 | 81 | |
BC013889.2 | - | 7 - 8 | 872 | 81 | |
KU178685.1 | - | 7 - 8 | 803 | 81 |