The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184976.1 | NP_001171905.1 | 6 - 7 | 551 | 70 |
NM_005337.4 | NP_005328.2 | 6 - 7 | 680 | 70 | |
GenBank mRNA | AK300783.1 | - | 6 - 7 | 551 | 70 |
BC093769.2 | - | 6 - 7 | 633 | 70 | |
BC093771.1 | - | 6 - 7 | 633 | 70 | |
M58285.1 | - | 6 - 7 | 641 | 70 |