The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002553.3 | NP_002544.1 | 7 - 8 | 880 | 143 |
NM_181747.3 | NP_859531.1 | 7 - 8 | 880 | 143 | |
GenBank mRNA | AF047599.1 | - | 7 - 8 | 815 | 143 |
AF049127.1 | - | 7 - 8 | 811 | 143 | |
AF081459.1 | - | 7 - 8 | 790 | 143 | |
AK223361.1 | - | 7 - 8 | 880 | 143 | |
AK292285.1 | - | 7 - 8 | 856 | 143 | |
AK302122.1 | - | 8 - 9 | 967 | 143 | |
AK313596.1 | - | 7 - 8 | 837 | 143 | |
BC023652.2 | - | 7 - 8 | 818 | 143 | |
U92538.1 | - | 7 - 8 | 825 | 143 |