The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206691.1 | NP_001193620.1 | 7 - 8 | 834 | 122 |
NM_032491.5 | NP_115880.2 | 3 - 4 | 500 | 122 | |
NM_213594.2 | NP_998759.1 | 7 - 8 | 1081 | 122 | |
GenBank mRNA | AB044245.1 | - | 7 - 8 | 799 | 122 |
AB095365.1 | - | 7 - 8 | 829 | 122 | |
AB095366.1 | - | 7 - 8 | 1081 | 122 | |
AF332192.1 | - | 3 - 4 | 494 | 122 | |
AK074913.1 | - | 4 - 5 | 412 | 122 | |
AK291445.1 | - | 7 - 8 | 1066 | 122 | |
AK315698.1 | - | 3 - 4 | 480 | 122 | |
AY102009.1 | - | 7 - 8 | 734 | 122 | |
BC028582.2 | - | 3 - 4 | 503 | 122 | |
BC030644.1 | - | 3 - 4 | 475 | 122 |