Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPH3A
Entrez Gene ID
22895
Gene Name
rabphilin 3A
Gene Aliases
-
Chromosome Location
Chr.12: 112791538 - 112898881 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143854.1 NP_001137326.1 6 - 7 775 74
NM_014954.3 NP_055769.2 5 - 6 763 74
XM_011538048.1 XP_011536350.1 6 - 7 579 74
GenBank mRNA AB023202.1 - 6 - 7 696 74
AK295696.1 - 6 - 7 729 74
AK296296.1 - 4 - 5 314 74
AK316112.1 - 6 - 7 610 74
BC002605.2 - 4 - 5 501 74
BC009843.2 - 4 - 5 424 74
BC017259.2 - 5 - 6 570 74

View Full Product Details