The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282460.1 | NP_001269389.1 | 3 - 4 | 1110 | 99 |
XM_011534989.2 | XP_011533291.1 | 3 - 4 | 1260 | 99 | |
XM_011534990.2 | XP_011533292.1 | 3 - 4 | 1260 | 99 | |
XM_011534992.2 | XP_011533294.1 | 3 - 4 | 1260 | 99 | |
XM_011534993.1 | XP_011533295.1 | 3 - 4 | 1260 | 99 | |
XM_017020421.1 | XP_016875910.1 | 3 - 4 | 1260 | 99 | |
XM_017020422.1 | XP_016875911.1 | 2 - 3 | 1016 | 99 | |
XM_017020423.1 | XP_016875912.1 | 2 - 3 | 1430 | 99 | |
XM_017020424.1 | XP_016875913.1 | 3 - 4 | 1260 | 99 | |
XM_017020425.1 | XP_016875914.1 | 3 - 4 | 1260 | 99 | |
XM_017020426.1 | XP_016875915.1 | 3 - 4 | 1260 | 99 | |
XM_017020427.1 | XP_016875916.1 | 3 - 4 | 1260 | 99 | |
XM_017020428.1 | XP_016875917.1 | 3 - 4 | 1134 | 99 | |
GenBank mRNA | BC030276.1 | - | 3 - 4 | 1105 | 99 |