The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007278.2 | NP_001007279.1 | 1 - 2 | 213 | 94 |
NM_052811.3 | NP_434698.1 | 1 - 2 | 213 | 94 | |
GenBank mRNA | AK314496.1 | - | 1 - 2 | 172 | 94 |
AY191002.1 | - | 1 - 2 | 161 | 94 | |
AY764035.1 | - | 1 - 2 | 183 | 94 | |
BC003579.1 | - | 1 - 2 | 174 | 94 |