Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP5J2
Entrez Gene ID
9551
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit F2
Gene Aliases
ATP5JL
Chromosome Location
Chr.7: 99458161 - 99466201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003714.2 NP_001003714.1 1 - 2 93 61
NM_004889.3 NP_004880.1 1 - 2 93 61
GenBank mRNA AF047436.1 - 1 - 2 60 61
AF088918.1 - 1 - 2 46 61
AK223348.1 - 1 - 2 56 61
BC003678.1 - 1 - 2 56 61
CB110074.1 - 1 - 2 60 61
CR456891.1 - 1 - 2 33 61
CR542155.1 - 1 - 2 33 61
DA064847.1 - 1 - 2 56 61
EL952520.1 - 1 - 2 47 61

View Full Product Details